Ac-D-Pro-OH
Glass bottle for solids, 1 g
Catalog # BACE4004420.0001
Marketsource
Supplier: BACHEM BIOCHEMICA
Ac-D-Pro-OH
Catalog # BACE4004420.0001
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 59785-68-1
Jump to:
Product Details & Documents
Synonyms: N-Acetyl-D-proline
- CAS Number:59785-68-1
- Formula:C₇H₁₁NO₃
- MDL Number:MFCD00080841
- Molecular Weight:157.17 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



