Isopentyl nitrite (mixed isomers ) 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-46-3
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00002057
:117.15 g/mol
Merck Index:14,5123
Boiling Pt:95…100 °C
Density:0.875
Flash Pt:–20 °C (–4 °F)
UN:1113
ADR:3,II
:Stabilised with sodium carbonate (Na₂CO₃) 0.2% (anhy)
EINECS:203-770-8
:GHS02
:GHS06
Storage Temperature:Refrigerator