

tert-Butylchlorodiphenylsilane 98%, AcroSeal®
430921000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Clear colorless to yellow to light brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5665 to 1.5685 (20°C, 589 nm)
- GC:≥97.5 %
:
:274.87 g/mol
:135 °C (.7 mmHg)
:1.06 g/cm³
:149 °C
:Ambient
:261-282-0
:2987
:8,II
:GHS07
:GHS05
:58479-61-1
:C₁₆H₁₉ClSi
:MFCD00000497