Order Entry
Germany
Orders LinkContactUsLinkComponent
 

N-Acetyl-L-proline 99%

N-Acetyl-L-proline 99%
Certificates
N-Acetyl-L-proline 99%

2 Options Available Below

Product Details & Documents
Synonyms:  N-Acetyl-L-Proline, Ac-Pro-OH
  • CAS Number:68-95-1
  • Formula:C₇H₁₁NO₃
  • MDL Number:MFCD00020837
  • Molecular Weight:157.169 g/mol
  • Melting Pt:115…117 °C
  • Storage Temperature:Refrigerator
  • EINECS:276-259-0

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Specific optical rotation:
    -114° to -118° (20°C, 589 nm) (C=1, Water)
  • Titration with TBAH:
    98.5 to 101.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 42,60 €
    42,60 €
    1 * 1 g
       42,60 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 179,00 €
    179,00 €
    1 * 5 g
       179,00 €