0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Trityl chloride
Trityl chloride ≥98%
Trityl chloride ≥98%
:
Thermo Fisher Scientific
Trityl chloride ≥98%
:
Thermo Fisher Scientific
:
76-83-5
:
:
C₁₉H₁₅Cl
:
MFCD00000813
:
278.78 g/mol
:
230…235 °C (20 mmHg)
:
110…114 °C
:
1.25 g/cm³ (20 °C)
:
Ambient
:
200-986-4
:
3261
:
8,II
:
GHS05
msds_ALFAA11799
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years