Syringes, SteriPlast®
Supplier: BURKLE
Certificates
About this item
Transparent PP syringes with printed graduations.
- Can be extended to any length with additional tubing
- Luer adapter is supplied for the 100 ml syringe
The syringe is supplied in a sterile package and ready for immediate use.