Propylbenzene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-65-1
:
Formula:C₉H₁₂
MDL Number:MFCD00009377
:120.19 g/mol
Merck Index:12,08029
Boiling Pt:159 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.86 g/cm³
Flash Pt:47 °C
EINECS:203-132-9
UN:2364
ADR:3,III
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4900 to 1.4930 (20°C, 589 nm)
- GC:≥97.5 %
- Specific optical rotation:+39° to +69° (20°C, 589 nm) (C=2, Chloroform)