Order Entry
Germany
ContactUsLinkComponent
Tapes, accession numbered
Tapes, accession numbered
  TIMECNR-10
undefined
Tapes, accession numbered
  TIMECNR-10

 

  • Colour:
    Black on white
  • Description:
    Accession numbered tape, custom imprinted
  • Dimensions:
    13 mm×19 m
  • Material:
    Paper
  • Pk:
    1.000