tert-Butylbenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-06-6
:
Formula:C₁₀H₁₄
MDL Number:MFCD00008816
:134.22 g/mol
Merck Index:13,01550
Boiling Pt:169 °C (760 mmHg)
Melting Pt:–58 °C
Density:0.86 g/cm³
Flash Pt:44 °C
Storage Temperature:Ambient
EINECS:202-632-4
UN:2709
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Water:≤0.5 % (K.F.)
- Color scale:≤10 APHA
- Residue after ignition:≤0.005 %
- Substance darkened by H2SO4:Passes test
- Sulfate (SO4):≤10 ppm
- Chlorinated cmpds (as Cl):≤0.003 %
- Heavy metals (as Pb):≤2 ppm
- Impurity:(Acrolein and glucose) Passes test
- Fatty acid esters:≤0.05 % (as Butyric acid)
- Neutrality:Passes test