Trolleys, basket, KR 60-60
Catalog # 139-0800
Supplier: HUPFER METALLWERKE
Specifications
- Description:Basket trolley with 4 swivel castors (2 of them with brakes), KR
- W×D×H:770×675×855 mm
- Pk:1
- Shelves:1
Specifications
About this item
These trolleys made of stainless steel can take baskets, containers and bins made from polypropylene.
- Stable, mobile round tube construction
- Mobile on four swivel castors, two of them with brake
- Galvanised-chromed housings with electro-conductive double ball-bearing
- Two bumpers protect trolley against damage