Orders LinkContactUsLinkComponent
Serological pipettes
Serological pipettes
Serological pipettes
Supplier:  RATIOLAB
Certificates
Serological pipettes
Serological pipettes
4 Options Available Below
Product Details & Documents
Disposable pipettes made from clear polystyrene (PS).
  • Free from RNase, DNase, DNA and pyrogens
  • Sterile, individually packed
  • From one mould – hence more accurate and stable and with minimal residual wetting
  • Permanent black embossed graduations
  • Colour coded cotton wool plug for convenient identification
  • Negative graduation for additional capacity
  • 5, 10 and 25 ml pipettes with scales for dispensed and the residual volumes
Product Family Options
Serological pipettes
Product Information
  • Capacity
    Division
    Sterile
    Colour code
    Packed
    Availability
    Price
  • 10 ml
    0,1 ml
    +
    Orange
    Individually
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 55,40 €
    55,40 €
    1 * 200 ST
       55,40 €
  • 25 ml
    0,2 ml
    +
    Red
    Individually
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 141,50 €
    141,50 €
    1 * 200 ST
       141,50 €
  • 50 ml
    0,5 ml
    +
    Purple
    Individually
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 166,70 €
    166,70 €
    1 * 100 ST
       166,70 €
  • 2 ml
    0,02 ml
    +
    Green
    Individually
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 153,50 €
    153,50 €
    1 * 1.000 ST
       153,50 €