Order Entry
Germany
ContactUsLinkComponent
Double bosshead, cross type
Double bosshead, cross type
Catalog # BOCH5311
Supplier:  BOCHEM
undefined
Double bosshead, cross type
Catalog # BOCH5311
Supplier:  BOCHEM

Specifications

  • Material:
    Stainless steel 18/10
  • Pk:
    1
  • For rod Ø:
    16,5 mm

Specifications

About this item

Cross type double bossheads made of stainless steel.

  • For rods up to 16,5 mm
  • Angle: 90 + 180°
  • With socket screw (M6)