Orders LinkContactUsLinkComponent
Safety Bottle Container
Safety Bottle Container
Supplier:   BOCHEM
Safety Bottle Container
Supplier:  BOCHEM
Certificates
Safety Bottle Container
Safety Bottle Container
4 Options Available Below
Product Details & Documents
Safety transport container, made of stainless steel, for laboratory bottles.
  • For bottles 50 to 1000 ml
  • With hinge, lock and silicone seal
  • Good chemical resistance
Product Family Options
Silicone Seals
Product Information
  • Capacity
    Colour
    For
    Ø ext.×H
    Availability
    Price
  • BOCH18301SS
    Silicone seal for transport container
    Silicone seal for transport container
    Red
    Container with Ø 160 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 23,20 €
    23,20 €
    1 * 5 ST
       23,20 €
  • BOCH18300SS
    Silicone seal for transport container
    Silicone seal for transport container
    Red
    Container with Ø 130 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 22,40 €
    22,40 €
    1 * 5 ST
       22,40 €
Product Information
  • Capacity
    Colour
    For
    Ø ext.×H
    Availability
    Price
  • 1 l
    Stainless steel
    50 - 250 ml flasks
    130×140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 323,00 €
    323,00 €
    1 * 1 ST
       323,00 €
  • 2 l
    Stainless steel
    500 - 1000 ml flasks
    160×250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 378,00 €
    378,00 €
    1 * 1 ST
       378,00 €