Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Isopropylaniline
2-Isopropylaniline
2-Isopropylaniline
Irritant
2-Isopropylaniline
Supplier:
CHEMPUR
Certificates
Expand image
Irritant
2-Isopropylaniline
Supplier:
CHEMPUR
About this item
CAS Number:
643-28-7
:
Formula:
C₉H₁₃N
MDL Number:
MFCD00007720
:
135.21 g/mol
EINECS:
211-397-7
:
GHS07