Silicone sealing rings with PTFE washer DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
VMQ with bonded PTFE face.
- Temperature resistant from −200 to +250 °C
- Very good chemical resistance
- Easy handling
For threaded connection caps with hole and GL thread.