Orders LinkContactUsLinkComponent
Mopping systems, EasyCurve™
Supplier:   CONTEC
Mopping systems, EasyCurve™
Supplier:  CONTEC
Certificates
Mopping systems, EasyCurve™
Mopping systems, EasyCurve™
5 Options Available Below
Product Details & Documents
Contec’s EasyCurve™ Mop was designed specifically for critical and controlled environments, now available with a ribbed microfiber face giving users smooth and easy mopping for better particle pick up and contamination removal. The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability, making the EasyCurve™ an ideal cleaning tool for ceilings, walls and floors. Available in nonsterile and validated sterile versions.
Mop heads:
Very absorbent and drip-free
Compatible with most common solutions, solvents and disinfectants
Better contaminant removal
Economical and easy to install without tools
Mop frame:
Compatible with Contec EasyCurve mops™ and QuickConnect™ handles
Mop head frame 2724 may be sterilized by most common methods including gamma irradiation, ethylene oxide, or autoclave (121 °C for 30 minutes)
Provides effortless contact with surfaces
Ensures complete and uniform contact with all surfaces
All components of the EasyCurve system are autoclavable making it an ideal upgrade alternative in applications where sponge roller and self-wringing style mops are currently used.
Product Family Options
Product Information
  • Dimensions
    Packed
    Availability
    Price
  • CONT2724
    Mop head frame, stainless steel, EasyCurve™
    Mop head frame, stainless steel, EasyCurve™
    360×130×70 mm
    1/case
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 189,00 €
    189,00 €
    1 * 1 ST
       189,00 €
  • CONTEZMP0350
    Sterile ribbed microfibre mop head foam/polyester ribbed microfibre mop, EasyCurve™
    Sterile ribbed microfibre mop head foam/polyester ribbed microfibre mop, EasyCurve™
    360×110×20 mm
    1/bag; 48 bags/case
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 1 340,00 €
    1 340,00 €
    1 * 48 ST
       1 340,00 €
  • CONTEZMP0300
    Ribbed microfibre mop head, EasyCurve™
    Ribbed microfibre mop head, EasyCurve™
    360×110×20 mm
    4/bag; 12 bags/case
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 1 040,00 €
    1 040,00 €
    1 * 48 ST
       1 040,00 €
  • CONTEZSE0150
    Sterile sealed edge mop head, EasyCurve™
    Sterile sealed edge mop head, EasyCurve™
    380×130×10 mm
    1/bag; 24 bags/case
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 500,00 €
    500,00 €
    1 * 24 ST
       500,00 €
  • CONTEZSE0100
    Sealed edge mop head, EasyCurve™
    Sealed edge mop head, EasyCurve™
    380×130×10 mm
    2/bag; 20 bags/case
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 749,00 €
    749,00 €
    1 * 40 ST
       749,00 €