Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
Certificates
About this item
Tubes made of PP, translucent, caps made of PE.
- Available in plain or graduated configuration
- Max. RCF: Non skirted tubes can be centrifuged up to 20000×g
- Temperature range: –196 to +110 °C
- Flat screw caps with lip seal