Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-5-nitrotoluene
2-Fluoro-5-nitrotoluene
2-Fluoro-5-nitrotoluene
2-Fluoro-5-nitrotoluene
Supplier:
FLUOROCHEM
Certificates
Expand image
2-Fluoro-5-nitrotoluene
Supplier:
FLUOROCHEM
About this item
CAS Number:
455-88-9
:
Formula:
C₇H₆FNO₂
MDL Number:
MFCD00007284
:
155.13 g/mol
Boiling Pt:
230 °C (1013 hPa)
Melting Pt:
40 °C
EINECS:
207-251-7
UN:
2811
ADR:
6.1,III