Order Entry
Germany
Orders LinkContactUsLinkComponent
Benzotrifluoride

Benzotrifluoride

Glass bottle for solids, 0,25 g
Catalog # EHERC17846000
Flammable
Environment
Supplier:   EHRENSTORFER
Benzotrifluoride
Catalog # EHERC17846000
Supplier:  EHRENSTORFER
CAS Number:  98-08-8
Benzotrifluoride
Benzotrifluoride

Glass bottle for solids, 0,25 g

CAS Number:  98-08-8
Price
Inquire For Price
Log in for availability
1 * 0,25 g
Product Details & Documents
Synonyms:  α,α,α-Trifluorotoluene, Solvent BTF
  • CAS Number:98-08-8
  • Formula:C₇H₅F₃
  • MDL Number:MFCD00000372
  • Molecular Weight:146.11 g/mol
  • Merck Index:13,01111
  • Boiling Pt:101 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.37 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:202-635-0
  • UN:2338
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g

Specifications