Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Chloro-2-methylanisole

4-Chloro-2-methylanisole

Glass bottle for solids, 10 g
Catalog # FLUO018391-10G
Marketsource
Supplier:   FLUOROCHEM
4-Chloro-2-methylanisole
Catalog # FLUO018391-10G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  3260-85-3
4-Chloro-2-methylanisole

Glass bottle for solids, 10 g

CAS Number:  3260-85-3
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1-Chloro-4-methoxy-3-methylbenzene, 5-Chloro-2-methoxytoluene, 4-Chloro-1-methoxy-2-methylbenzene
  • CAS Number:3260-85-3
  • Formula:C₈H₉ClO
  • MDL Number:MFCD00070771
  • Molecular Weight:156.61 g/mol
  • Boiling Pt:212…214 °C (1013 hPa)
  • Melting Pt:37 °C
  • EINECS:221-860-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications