Pesticide reference standard, 2,4,5-Trichlorotoluene 10 µg/ml in hexane, Dr Ehrenstorfer
EHERLA17788000HE
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:6639-30-1
- Description:2,4,5-Trichlorotoluene 10 µg/ml in hexane
- Formula:C₇H₅Cl₃
- Description:2,4,5-Trichlorotoluene