α-Methylstyrene stabilised for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:98-83-9
:
Formula:C₉H₁₀
MDL Number:MFCD00008859
:118.18 g/mol
Boiling Pt:165.4 °C (1013 hPa)
Melting Pt:–23 °C
Density:0.9108 g/cm³ (20 °C)
Flash Pt:43 °C
EINECS:202-705-0
UN:2303
ADR:3,III
:M-Clarity™ quality level = MQ200
:Stabilised with 4-tert-butylpyrocatechol (TBC)
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:167 g/l (48 °C)
- Molar Mass:99.17 g/mol
- Boiling Point:105 - 108 °C
- Vapor Pressure:41 hPa (20 °C)
- Flash Point:3 °C
- Refractive Index:1.4378 (20 °C, 589 nm)
- Explosion Limit:1.1 - 9.9 %(V)
- Density:0.82 g/cm3 (20 °C)
- pH:12 (100 g/l, H2O, 20 °C)
- Ignition Point:205 °C