ContactUsLinkComponent
Sealing mats for 96-well PCR plates, Amplate™
Sealing mats for 96-well PCR plates, Amplate™
Catalog # SIMPT329-10
Supplier:  Simport Scientific
undefined
Sealing mats for 96-well PCR plates, Amplate™
Catalog # SIMPT329-10
Supplier:  Simport Scientific
Specifications
  • Colour:
    Blue
  • Description:
    Sealing mat for 96-well PCR plates
  • Material:
    Santoprene™
  • Pk:
    5