Order Entry
Germany
ContactUsLinkComponent
3-Methyl-1-phenyl-2-butanone 96%
3-Methyl-1-phenyl-2-butanone 96%
Catalog # L07176.06
CAS Number:  
3-Methyl-1-phenyl-2-butanone 96%
Catalog # L07176.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :162.23 g/mol
Boiling Pt:237…240 °C
Density:0.968
Storage Temperature:Ambient
EINECS:220-765-6
CAS Number:2893-05-2
Formula:(C₆H₅)CH₂C(O)CH(CH₃)CH₃
MDL Number:MFCD00040487