PCR plates, 96-well, opaque, skirted, Amplate™
Supplier: Simport Scientific
Certificates
About this item
PP. These opaque 96-well PCR plates are for chemiluminescent and fluorescent procedures. Thin-walled and designed for rapid thermal transfer, these plates are precision-moulded to ensure well-to-well and plate-to-plate uniformity.
- Opaque to ensure low level of background fluorescence
- Alphanumeric grid helps sample identification
- Corner at H1 of the plate is cut away to facilitate orientation
- Easy to seal since no cylindrical walls extend above the plate
- Area for bar coding, labelling or writing
Can be handled by robotic handling equipment and is ideal with automated pipetting systems.
Delivery: Packed in tamperproof resealable bags of 10 plates.