Orders LinkContactUsLinkComponent
Support clamps
Support clamps
Supplier:   BOCHEM
Support clamps
Supplier:  BOCHEM
Certificates
Support clamps
7 Options Available Below
Product Details & Documents
Support clamps made of stainless steel (18/19) and PTFE for either Schott laboratory flat flanges (PF) (optimal flange angle 45°) or Schott "Kugel-Flanges" (ball and socket system) (KF).
  • For reactors and other vessels
  • User friendly thanks to joint screws
  • With two supporting devices for 26,9 mm tubes
Product Family Options
Product Information
  • For
    Availability
    Price
  • DN 60
    Schott laboratory flat flanges (PF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 887,00 €
    887,00 €
    1 * 1 ST
       887,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17210
  • DN 100
    Schott laboratory flat flanges (PF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 894,00 €
    894,00 €
    1 * 1 ST
       894,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17211
  • DN 120
    Schott laboratory flat flanges (PF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 944,00 €
    944,00 €
    1 * 1 ST
       944,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17212
  • DN 150
    Schott laboratory flat flanges (PF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17213
  • DN 200
    Schott laboratory flat flanges (PF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 090,00 €
    1 090,00 €
    1 * 1 ST
       1 090,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17214
  • DN 100
    Schott "Kugel-Flanges" (KF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 925,00 €
    925,00 €
    1 * 1 ST
       925,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17215
  • DN 150
    Schott "Kugel-Flanges" (KF)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BOCH17216