Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methylpent-1-en-4-yn-3-ol
3-Methylpent-1-en-4-yn-3-ol, Sigma-Aldrich®
3-Methylpent-1-en-4-yn-3-ol, Sigma-Aldrich®
3-Methylpent-1-en-4-yn-3-ol, Sigma-Aldrich®
SIAL493023-5G
:
Merck
:
3-Methylpent-1-en-4-yn-3-ol, Sigma-Aldrich®
SIAL493023-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
3,III
:
M-Clarity™ quality level = MQ100
:
96.13 g/mol
:
221-770-6
:
1987
:
3230-69-1
:
C₆H₈O
:
MFCD01709006