Triisopropylamine
Catalog # FLUO020872-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:143.27 g/mol
EINECS:222-317-5
UN:1993
ADR:3,II
CAS Number:3424-21-3
Formula:C₉H₂₁N