2,3-Dimethoxy-5-pyridineboronic acid pinacol ester
Catalog # COBBPN-5388-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.12 g/mol
CAS Number:1083168-92-6
Formula:C₁₃H₂₀BNO₄
MDL Number:MFCD11857651