Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxyphenylboronic acid pinacol ester
4-Hydroxyphenylboronic acid pinacol ester
4-Hydroxyphenylboronic acid pinacol ester
4-Hydroxyphenylboronic acid pinacol ester
Supplier:
COMBI-BLOCKS
Certificates
Expand image
4-Hydroxyphenylboronic acid pinacol ester
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
269409-70-3
:
Formula:
C₁₂H₁₇BO₃
MDL Number:
MFCD02093756
:
220.08 g/mol