2,4-Dibutoxyphenylboronic acid
Catalog # COBBBB-8093-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.14 g/mol
CAS Number:870778-89-5
Formula:C₁₄H₂₃BO₄
MDL Number:MFCD07784348