Orders LinkContactUsLinkComponent
Clamps, universal
Clamps, universal
Supplier:   BOCHEM
Clamps, universal
Supplier:  BOCHEM
Certificates
Clamps, universal
Clamps, universal
2 Options Available Below
Product Details & Documents
These clamps made of stainless steel 18/10 feature cork lined fingers.
Certifications: DIN 12894
Product Family Options
Product Information
  • Spanning width
    Length
    Availability
    Price
  • BOCH5425
    Clamps, without cork lined jaws
    Clamps, without cork lined jaws
    0 - 80 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • BOCH5425-OK
    Clamps, without cork lined jaws
    Clamps, without cork lined jaws
    0 - 80 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 180,00 €
    180,00 €
    1 * 1 ST
       180,00 €