Spirit lamp, alcohol burner, stainless steel
451-0096
: BOCHEM
- Description:Spirit lamp, height 65 mm, Ø 85 mm
- Flask capacity:150 ml
- Pk:1
Spirit lamp made of 18/10 stainless steel.
- Non-magnetic
- Adjustable wick and flame
: Supplied as a complete set with burner, lid and cotton wick.