Order Entry
Germany
ContactUsLinkComponent
Microscope cover, acrylic, 216
Microscope cover, acrylic, 216
  630-9281
 :  KARTELL
undefined
Microscope cover, acrylic, 216
  630-9281
 :  KARTELL

 

  • Colour:
    Clear
  • Description:
    Acrylic microscope cover / bell, for 40 cm microscopes
  • W×D×H:
    300×307×400 mm
  • Pk:
    1

 

 

Clear acrylic resin microscope cover protects against dust and dirt.

  • Square base
  • For 40 cm microscopes

Size: 300x307x400 mm