Sodium hydrogen sulphate
Catalog # OVER813075-25KG
Supplier: OVERLACK CHEMISCHE PRODUKTE
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
Merck Index:12,08729
Melting Pt:181.85 °C
Density:1.8 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,III
:120.06 g/mol
EINECS:231-665-7
UN:3260
:GHS05
CAS Number:7681-38-1
Formula:HNaO₄S
MDL Number:MFCD00003531