Order Entry
Germany
ContactUsLinkComponent
Sodium chloride, Supelco®
Sodium chloride, Supelco®
Catalog # SIALZ123595-1EA
Supplier:  Merck
CAS Number:  
Sodium chloride, Supelco®
Catalog # SIALZ123595-1EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477