Order Entry
Germany
Orders LinkContactUsLinkComponent
Creatinine

Creatinine

Glass bottle for solids, 100 g
Catalog # MOLEM25285847
Marketsource
Supplier:   Molekula
Creatinine
Catalog # MOLEM25285847
Supplier:  Molekula
Marketsource
CAS Number:  60-27-5
Creatinine

Glass bottle for solids, 100 g

CAS Number:  60-27-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • EINECS:200-466-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications