Order Entry
Germany
Orders LinkContactUsLinkComponent
Creatinine 98%

Creatinine 98%

Creatinine 98%
Certificates
Creatinine 98%

3 Options Available Below

Product Details & Documents
Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Melting Pt:255 °C (dec.)
  • Flash Pt:290 °C
  • Storage Temperature:Ambient
  • EINECS:200-466-7

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellowish-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Sulfated ash:
    ≤0.2 %
  • Solubility:
    (5 % in water) Clear colorless to slightly yellow
  • Titration with HClO4:
    97.5 to 102.5 % (on dried substance)
  • Heavy metals (as Pb):
    ≤20 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.