Creatinine
1 KIT
Catalog # GRNB11711704
Supplier: GREINER BIOCHEMICAL
Jump to:
Product Details & Documents
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- EINECS:200-466-7
Specifications
Specifications
- Pk:1 KIT



