
1-(4-Morpholino)cyclohexene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:670-80-4
:
Formula:C₁₀H₁₇NO
MDL Number:MFCD00006163
:167.25 g/mol
Boiling Pt:118…120 °C (10 mmHg)
Density:1.020
Flash Pt:68 °C (154 °F)
EINECS:211-579-6
:GHS07