Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Aminophenylboronic acid pinacol ester
3-Aminophenylboronic acid pinacol ester
3-Aminophenylboronic acid pinacol ester
3-Aminophenylboronic acid pinacol ester
Supplier:
FLUOROCHEM
Certificates
Expand image
3-Aminophenylboronic acid pinacol ester
Supplier:
FLUOROCHEM
About this item
CAS Number:
210907-84-9
:
Formula:
C₁₂H₁₈BNO₂
MDL Number:
MFCD03453668
:
219.09 g/mol