Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminophenylboronic acid pinacol ester
2-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®
2-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®
2-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2-Aminophenylboronic acid pinacol ester, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
191171-55-8
:
Formula:
C₁₂H₁₈BNO₂
MDL Number:
MFCD02179448
:
219.09 g/mol
:
M-Clarity™ quality level = MQ100