Ethyl isothiocyanatoformate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16182-04-0
:
Formula:C₄H₅NO₂S
MDL Number:MFCD00004814
:131.16 g/mol
Boiling Pt:44…46 °C (10 mmHg)
Density:1.110
Flash Pt:50 °C (122 °F)
EINECS:240-318-9
UN:2929
ADR:6.1,II
:GHS06
:GHS08
:GHS02
:GHS07