Order Entry
Germany
ContactUsLinkComponent
4'-Hydroxybiphenyl-4-carboxylic acid 99%, Sigma-Aldrich®
4'-Hydroxybiphenyl-4-carboxylic acid 99%, Sigma-Aldrich®
Catalog # 371335-5G
CAS Number:  
undefined
4'-Hydroxybiphenyl-4-carboxylic acid 99%, Sigma-Aldrich®
Catalog # 371335-5G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance (colour):
    White to Light Brown
  • Appearance (form):
    Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
  • Titration (T) NaOH 0.1 m:
    98.5 - 101.5%
  • Purity (HPLC area%):
    ≥ 98.5%
  • Infrared spectrum:
    Conforms to structure

Specification Test Results

About this item

 :
 :214.22 g/mol
Melting Pt:295 °C (dec., lit.)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
CAS Number:58574-03-1
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00059078