4'-Methoxybiphenyl-2-carboxylic acid
Catalog # FLUO011466-500MG
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:228.25 g/mol
CAS Number:18110-71-9
Formula:C₁₄H₁₂O₃
MDL Number:MFCD03426469