Order Entry
Germany
Orders LinkContactUsLinkComponent
(E)-β-Ionone

(E)-β-Ionone

Glass bottle for solids, 100 g
Catalog # MOLEM35676152
Marketsource
Supplier:   Molekula
(E)-β-Ionone
Catalog # MOLEM35676152
Supplier:  Molekula
Marketsource
CAS Number:  79-77-6
(E)-β-Ionone

Glass bottle for solids, 100 g

CAS Number:  79-77-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone
  • CAS Number:79-77-6
  • Formula:C₁₃H₂₀O
  • MDL Number:MFCD00001549
  • Molecular Weight:192.30 g/mol
  • EINECS:201-224-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications