Order Entry
Germany
Orders LinkContactUsLinkComponent
 

(S)-(+)-2-Hydroxyisovaleric acid

Supplier:   Cayman Chemical
(S)-(+)-2-Hydroxyisovaleric acid
Supplier:  Cayman Chemical
Certificates
(S)-(+)-2-Hydroxyisovaleric acid

3 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
  • CAS Number:17407-55-5
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066443
  • Molecular Weight:118.13 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 46,00 €
    46,00 €
    1 * 500 mg
       46,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 78,00 €
    78,00 €
    1 * 1 g
       78,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 280,00 €
    280,00 €
    1 * 5 g
       280,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.