


Isophorone diisocyanate (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4098-71-9
:
Formula:C₁₂H₁₈N₂O₂
MDL Number:MFCD00064956
:222.29 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:223-861-6
UN:2290
:GHS06
:GHS08
:GHS09
Specification Test Results
- Solubility:0.015 g/l (20 °C) (slow decomposition)
- Melting Point:-60 °C
- Molar Mass:222.29 g/mol
- Boiling Point:158 - 159 °C (13 hPa)
- Vapor Pressure:0.0004 hPa (20 °C)
- Flash Point:155 °C
- Refractive Index:1.482 (25 °C, 589 nm)
- Explosion Limit:0.7 - 4.5 %(V)
- Density:1.06 g/cm3 (20 °C)
- Ignition Point:430 °C
- Synonyms:5-Isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-Isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate
- Hill Formula:C12H18N2O2