


Isophorone diisocyanate (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4098-71-9
:
Formula:C₁₂H₁₈N₂O₂
MDL Number:MFCD00064956
:222.29 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:223-861-6
UN:2290
:GHS06
:GHS08
:GHS09
Frequently Bought Together