2-(Trimethylsilyl)thiazole 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79265-30-8
:
Formula:C₆H₁₁NSSi
MDL Number:MFCD00066274
:157.31 g/mol
Boiling Pt:53…55 °C (10 mmHg)
Density:0.98 g/cm³
Flash Pt:62 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.5000 (20°C, 589 nm)
- GC:≥95 %